4-Iodo-2-nitroanisole

Supplier: Apollo Scientific

Synonyms: 4-Iodo-1-methoxy-2-nitrobenzene

OR400184-5G
APOSOR400184-5GEA 97.3 EUR
APOSOR400184-5G
4-Iodo-2-nitroanisole
4-Iodo-2-nitroanisole
Formula: C₇H₆INO₃
MW: 279.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08060933
CAS Number: 52692-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR