6-Chloro-5-nitroquinoline 98%

Supplier: Apollo Scientific

OR41004-5G
APOSOR41004-5GEA 79.9 EUR
APOSOR41004-5G
6-Chloro-5-nitroquinoline 98%
6-Chloro-5-nitroquinoline
Formula: C₉H₅ClN₂O₂
MW: 208.6012 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00093616
CAS Number: 86984-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR