4-Amino-6-iodoindole 98%

Supplier: Apollo Scientific

Synonyms: 6-Iodo-1H-indol-4-amine

OR450013-250MG
APOSOR450013-250MGEA 260 EUR
APOSOR450013-250MG
4-Amino-6-iodoindole 98%
4-Amino-6-iodoindole
Formula: C₈H₇IN₂
MW: 258.059 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781458
CAS Number: 885520-58-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR