4-Phenyltetrahydropyran-4-ylamine

Supplier: Apollo Scientific

OR451049-1G
APOSOR451049-1GEA 1090 EUR
APOSOR451049-1G
4-Phenyltetrahydropyran-4-ylamine
4-Phenyltetrahydropyran-4-ylamine
Formula: C₁₁H₁₅NO
MW: 177.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09903681
CAS Number: 14006-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR