4-Iodo-2-methoxyphenol

Supplier: Apollo Scientific
Danger

Synonyms: 4-Iodoguaiacol, 2-Hydroxy-5-iodoanisole

OR46089-1G OR46089-25G OR46089-5G
APOSOR46089-1GEA 56.7 EUR
APOSOR46089-1G APOSOR46089-25G APOSOR46089-5G
4-Iodo-2-methoxyphenol
4-Iodo-2-methoxyphenol
Formula: C₇H₇IO₂
MW: 250.034 g/mol
Melting Pt: 37…40 °C
Storage Temperature: Ambient
MDL Number: MFCD08061924
CAS Number: 203861-62-5
UN: 2811
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR