1-(4-Methylpyridin-3-yl)ethanone

Supplier: Apollo Scientific
Warning

Synonyms: 1-(4-Methyl-3-pyridinyl)-Ethanone

OR46708-1G OR46708-250MG OR46708-500MG
APOSOR46708-1GEA 91.5 EUR
APOSOR46708-1G APOSOR46708-250MG APOSOR46708-500MG
1-(4-Methylpyridin-3-yl)ethanone
1-(4-Methylpyridin-3-yl)ethanone
Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 57…58 °C
Storage Temperature: Ambient
MDL Number: MFCD08447082
CAS Number: 51227-30-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR