3-(4-Methyl-1-piperazinylsulphonyl)benzoic acid

Supplier: Apollo Scientific

OR470025-5G
APOSOR470025-5GEA 519 EUR
APOSOR470025-5G
3-(4-Methyl-1-piperazinylsulphonyl)benzoic acid
3-(4-Methyl-1-piperazinylsulphonyl)benzoic acid
Formula: C₁₂H₁₆N₂O₄S
MW: 284.331 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02725037
CAS Number: 380339-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR