4-Isopropoxysalicylaldehyde

Supplier: Apollo Scientific

OR470744-5G
APOSOR470744-5GEA 916 EUR
APOSOR470744-5G
4-Isopropoxysalicylaldehyde
4-Isopropoxysalicylaldehyde
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08753660
CAS Number: 52085-11-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR