8-Cbz-1,8-diazaspiro[5.5]undecane

Supplier: Apollo Scientific

OR470845-1G
APOSOR470845-1GEA 1420 EUR
APOSOR470845-1G
8-Cbz-1,8-diazaspiro[5.5]undecane
8-Cbz-1,8-diazaspiro[5.5]undecane
Formula: C₁₇H₂₄N₂O₂
MW: 288.385 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11223536
CAS Number: 1086394-89-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR