2-[4-(2-Aminoethyl)phenoxy]ethanol

Supplier: Apollo Scientific

OR470901-1G
APOSOR470901-1GEA 916 EUR
APOSOR470901-1G
2-[4-(2-Aminoethyl)phenoxy]ethanol
2-[4-(2-Aminoethyl)phenoxy]ethanol
Formula: C₁₀H₁₅NO₂
MW: 181.232 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08449030
CAS Number: 4960-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR