2-[(1,2,4-Triazol-1-yl)methyl]aniline

Supplier: Apollo Scientific

OR470961-5G
APOSOR470961-5GEA 601 EUR
APOSOR470961-5G
2-[(1,2,4-Triazol-1-yl)methyl]aniline
2-[(1,2,4-Triazol-1-yl)methyl]aniline
Formula: C₉H₁₀N₄
MW: 174.202 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09025845
CAS Number: 127988-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR