Ethyl 3-(2-Cyano-N-methylacetamido)propanoate

Supplier: Apollo Scientific

OR471079-1G
APOSOR471079-1GEA 1120 EUR
APOSOR471079-1G
Ethyl 3-(2-Cyano-N-methylacetamido)propanoate
Ethyl 3-(2-Cyano-N-methylacetamido)propanoate
Formula: C₉H₁₄N₂O₃
MW: 198.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16147409
CAS Number: 1306948-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR