3-Hydroxy-1-propylpiperidine

Supplier: Apollo Scientific

OR471449-1G
APOSOR471449-1GEA 519 EUR
APOSOR471449-1G
3-Hydroxy-1-propylpiperidine
3-Hydroxy-1-propylpiperidine
Formula: C₈H₁₇NO
MW: 143.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16153145
CAS Number: 27361-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR