1-(5-Chloroquinolin-8-yl)ethanone

Supplier: Apollo Scientific
Warning

OR471594-250MG
APOSOR471594-250MGEA 246 EUR
APOSOR471594-250MG
1-(5-Chloroquinolin-8-yl)ethanone
1-(5-Chloroquinolin-8-yl)ethanone
Formula: C₁₁H₈ClNO
MW: 205.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28338884
CAS Number: 1438559-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR