8-Methylquinoline 97%

Supplier: Apollo Scientific

OR480642-100G
APOSOR480642-100GEA 215 EUR
APOSOR480642-100G
8-Methylquinoline 97%
8-Methylquinoline
Formula: C₁₀H₉N
MW: 143.19 g/mol
Boiling Pt: 248
Melting Pt: -80
Storage Temperature: Ambient
MDL Number: MFCD00006810
CAS Number: 611-32-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR