Ethyl-4-cyano-5-(methylthio)thiophene-2-carboxylate 97%

Supplier: Apollo Scientific

Synonyms: 4-Cyano-5-methylsulphanylthiophene-2-carboxylic acid ethyl ester, Ethyl 4-cyano-5-(methylthio)thiophene-2-carboxylate

OR480788-1G OR480788-5G
APOSOR480788-1GEA 85.7 EUR
APOSOR480788-1G APOSOR480788-5G
Ethyl-4-cyano-5-(methylthio)thiophene-2-carboxylate 97%
Ethyl-4-cyano-5-(methylthio)thiophene-2-carboxylate
Formula: C₉H₉NO₂S₂
MW: 227.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01859680
CAS Number: 116170-84-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR