3-Bromotoluene 99%

Supplier: Apollo Scientific
Warning

OR4938-100G OR4938-25G OR4938-500G
APOSOR4938-100GEA 75.4 EUR
APOSOR4938-100G APOSOR4938-25G APOSOR4938-500G
3-Bromotoluene 99%
3-Bromotoluene
Formula: C₇H₇Br
MW: 171.04 g/mol
Boiling Pt: 182…184 °C
Melting Pt: –40 °C
Density: 1.41 g/cm³ (25 °C)
Flash Pt: 59 °C
Storage Temperature: Ambient
MDL Number: MFCD00000085
CAS Number: 591-17-3
EINECS: 209-702-3
UN: 1993
ADR: 3 III
Merck Index: 13,01424

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR