7-Bromo-3-chloro-4H-pyrido[1,2-a]pyrimidin-4-one ≥98%

Supplier: Apollo Scientific

OR510233-1G
APOSOR510233-1GEA 358 EUR
APOSOR510233-1G
7-Bromo-3-chloro-4H-pyrido[1,2-a]pyrimidin-4-one ≥98%
7-Bromo-3-chloro-4H-pyrido[1,2-a]pyrimidin-4-one
Formula: C₈H₄BrClN₂O
MW: 259.487 g/mol
Storage Temperature: Ambient
CAS Number: 1785021-08-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR