6-(1-Pyrrolidinyl)-3-pyridinamine

Supplier: Apollo Scientific

Synonyms: 5-Amino-2-(pyrrolidin-1-yl)pyridine

OR52050-1G OR52050-100MG OR52050-250MG OR52050-500MG OR52050-5G
APOSOR52050-1GEA 275 EUR
APOSOR52050-1G APOSOR52050-100MG APOSOR52050-250MG APOSOR52050-500MG APOSOR52050-5G
6-(1-Pyrrolidinyl)-3-pyridinamine
6-(1-Pyrrolidinyl)-3-pyridinamine
Formula: C₉H₁₃N₃
MW: 163.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03425745
CAS Number: 92808-19-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR