4,4'-Bis(2-benzoxazolyl)stilbene

Supplier: Apollo Scientific
Warning

Synonyms: 1,2-Bis(4-(benzo[d]oxazol-2-yl)phenyl)ethene

OR52140-100G OR52140-1KG OR52140-500G
APOSOR52140-100GEA 23.2 EUR
APOSOR52140-100G APOSOR52140-1KG APOSOR52140-500G
4,4'-Bis(2-benzoxazolyl)stilbene
4,4'-Bis(2-benzoxazolyl)stilbene

Optical brightener for plastics (PP, ABS, EVA, PS and PC) and fibre (polyester fibres and PP fibres).

Formula: C₂₈H₁₈N₂O₂
MW: 414.46 g/mol
Melting Pt: 353…359 °C
Storage Temperature: Ambient
MDL Number: MFCD00022846
CAS Number: 1533-45-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR