6-Chloro-4-methyl-3-nitropyridin-2-amine 95+%

Supplier: Apollo Scientific

OR52243-250MG
APOSOR52243-250MGEA 102 EUR
APOSOR52243-250MG
6-Chloro-4-methyl-3-nitropyridin-2-amine 95+%
6-Chloro-4-methyl-3-nitropyridin-2-amine
Formula: C₆H₆ClN₃O₂
MW: 187.584 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17016063
CAS Number: 863878-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR