6-Morpholinopyridin-2-amine 95%

Supplier: Apollo Scientific

OR52776-250MG
APOSOR52776-250MGEA 609 EUR
APOSOR52776-250MG
6-Morpholinopyridin-2-amine 95%
6-Morpholinopyridin-2-amine
Formula: C₉H₁₃N₃O
MW: 179.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18483100
CAS Number: 400774-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR