5-Cyano-2-methylbenzoic acid

Supplier: Apollo Scientific

OR53221-1G
APOSOR53221-1GEA 267 EUR
APOSOR53221-1G
5-Cyano-2-methylbenzoic acid
5-Cyano-2-methylbenzoic acid
Formula: C₉H₇NO₂
MW: 161.157 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13189533
CAS Number: 1975-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR