(2S)-2-(Methylamino)propan-1-ol 95%

Supplier: Apollo Scientific
Danger

OR55461-100MG OR55461-1G OR55461-250MG
APOSOR55461-100MGEA 139 EUR
APOSOR55461-100MG APOSOR55461-1G APOSOR55461-250MG
(2S)-2-(Methylamino)propan-1-ol 95%
(2S)-2-(Methylamino)propan-1-ol
Formula: C₄H₁₁NO
MW: 89.136 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07367992
CAS Number: 40916-73-2
UN: 2735
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR