3-Toluenethiol 98%

Supplier: Apollo Scientific
Danger

Synonyms: m-Thiocresol, 3-Methylbenzene-1-thiol, 3-Thiocresol, 3-Methylbenzenethiol, m-Toluenethiol, 1-Mercapto-3-methylbenzene, 3-Methylthiophenol

OR5559-5G
APOSOR5559-5GEA 58 EUR
APOSOR5559-5G
3-Toluenethiol 98%
3-Toluenethiol
Formula: C₇H₈S
MW: 124.21 g/mol
Boiling Pt: 196 °C
Density: 1.044 g/cm³ (25 °C)
Flash Pt: 73 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004843
CAS Number: 108-40-7
EINECS: 203-578-4
UN: 3082
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR