5-Methylfuran-2-sulphonyl chloride 95%

Supplier: Apollo Scientific
Danger

OR59996-1G OR59996-250MG
APOSOR59996-1GEA 764 EUR
APOSOR59996-1G APOSOR59996-250MG
5-Methylfuran-2-sulphonyl chloride 95%
5-Methylfuran-2-sulphonyl chloride
Formula: C₅H₅ClO₃S
MW: 180.61 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11109334
CAS Number: 69815-95-8
UN: 3265
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR