1,5-Diaminonaphthalene

Supplier: Apollo Scientific
Warning

Synonyms: 1,5-naphthylenediamine, 1,5-Naphthalenediamine

OR60078-5G
APOSOR60078-5GEA 21.9 EUR
APOSOR60078-5G
1,5-Diaminonaphthalene
1,5-Diaminonaphthalene
Formula: C₁₀H₁₀N₂
MW: 158.2 g/mol
Boiling Pt: 210 °C/5.25mmHg
Melting Pt: 185…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00004029
CAS Number: 2243-62-1
EINECS: 218-817-8
UN: 3077
ADR: 9+EHS III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR