Farnesol (mixture of isomers) 95%
Supplier: Apollo Scientific
Warning
Synonyms: Farnesol, 3,7,11-Trimethyldodeca-2,6,10-trien-1-ol, TRANS,TRANS-3,7,11-TRIMETHYL-2,6,10-DODECATRIEN-1-OL, 2,6,10-Dodecatrien-1-ol, 3,7,11-trimethyl-, (2Z,6Z)-
OR6039-100G
APOSOR6039-100GEA 106 EUR
APOSOR6039-100G
Farnesol (mixture of isomers) 95%
Farnesol (mixture of isomers)
Formula: C₁₅H₂₆O
MW: 222.37 g/mol
Boiling Pt: 149 °C/4mmHg
Density: 0.886 g/cm³ (25 °C)
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00002918
CAS Number: 4602-84-0
UN: 3082
ADR: 9+EHS III