[1,3]Thiazolo[5,4-b]pyridine

Supplier: Apollo Scientific
Warning

Synonyms: Thiazolo[5,4-b]pyridine

OR61213-1G
APOSOR61213-1GEA 256 EUR
APOSOR61213-1G
[1,3]Thiazolo[5,4-b]pyridine
[1,3]Thiazolo[5,4-b]pyridine
Formula: C₆H₄N₂S
MW: 136.18 g/mol
Melting Pt: 52…53 °C
Storage Temperature: Ambient
MDL Number: MFCD11840312
CAS Number: 273-84-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR