6-Amino-4-hydroxy-2-mercaptopyrimidine 97%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Amino-2-thioxo-2,3-dihydropyrimidin-4(1H)-one, 6-Aminothiouracil, 6-Amino-2-thiouracil

OR6128-100G OR6128-250MG OR6128-5G OR6128-10G OR6128-25G OR6128-50G
APOSOR6128-100GEA 116 EUR
APOSOR6128-100G APOSOR6128-250MG APOSOR6128-5G APOSOR6128-10G APOSOR6128-25G APOSOR6128-50G
6-Amino-4-hydroxy-2-mercaptopyrimidine 97%
6-Amino-4-hydroxy-2-mercaptopyrimidine
Formula: C₄H₅N₃OS
MW: 143.1670 g/mol
Melting Pt: 330…332 °C
Storage Temperature: Ambient
MDL Number: MFCD00006077
CAS Number: 1004-40-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR