4-Chloro-5-methylpyrimidin-2-amine 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2-Amino-4-chloro-5-methylpyrimidine

OR61403-1G OR61403-5G
APOSOR61403-1GEA 82.8 EUR
APOSOR61403-1G APOSOR61403-5G
4-Chloro-5-methylpyrimidin-2-amine 98%
4-Chloro-5-methylpyrimidin-2-amine
Formula: C₅H₆ClN₃
MW: 143.58 g/mol
Melting Pt: 184…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00725898
CAS Number: 20090-58-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR