4-Phenyl-2-pyridine carboxylic acid

Supplier: Apollo Scientific
Warning

OR6247-1G OR6247-50MG OR6247-500MG OR6247-5G
APOSOR6247-1GEA 380 EUR
APOSOR6247-1G APOSOR6247-50MG APOSOR6247-500MG APOSOR6247-5G
4-Phenyl-2-pyridine carboxylic acid
4-Phenyl-2-pyridine carboxylic acid
Formula: C₁₂H₉NO₂
MW: 199.21 g/mol
Melting Pt: 154…160 °C
Storage Temperature: Ambient
MDL Number: MFCD04114164
CAS Number: 52565-56-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR