Picolinoyl chloride hydrochloride 93%

Supplier: Apollo Scientific
Danger

Synonyms: Pyridine-2-carbonyl chloride hydrochloride

OR6370-1G OR6370-5G OR6370-10G
APOSOR6370-1GEA 21.9 EUR
APOSOR6370-1G APOSOR6370-5G APOSOR6370-10G
Picolinoyl chloride hydrochloride 93%
Picolinoyl chloride hydrochloride
Formula: C₆H₅Cl₂NO
MW: 178.0160 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00060193
CAS Number: 39901-94-5
UN: 3261
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR