5-Chloro-1-pentanol 95%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Chloropentan-1-ol

OR6509-5G OR6509-25G OR6509-100G
APOSOR6509-5GEA 21.9 EUR
APOSOR6509-5G APOSOR6509-25G APOSOR6509-100G
5-Chloro-1-pentanol 95%
5-Chloro-1-pentanol
Formula: C₅H₁₁ClO
MW: 122.59 g/mol
Boiling Pt: 102…103 °C/8mmHg
Density: 1.06 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00039556
CAS Number: 5259-98-3
EINECS: 226-067-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR