2-(3-Hydroxypropyl)benzimidazole 97%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(2-Benzimidazolyl)propanol, 3-(1H-Benzo[d]imidazol-2-yl)propan-1-ol, 2-Benzimidazolepropanol

OR6551-10G OR6551-25G OR6551-50G OR6551-5G OR6551-250MG OR6551-500MG
APOSOR6551-10GEA 379 EUR
APOSOR6551-10G APOSOR6551-25G APOSOR6551-50G APOSOR6551-5G APOSOR6551-250MG APOSOR6551-500MG
2-(3-Hydroxypropyl)benzimidazole 97%
2-(3-Hydroxypropyl)benzimidazole
Formula: C₁₀H₁₂N₂O
MW: 176.22 g/mol
Melting Pt: 164…166 °C
Storage Temperature: Ambient
MDL Number: MFCD00022688
CAS Number: 2403-66-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR