5-Bromo-2-methoxybenzoic acid

Supplier: Apollo Scientific
Danger

OR70056-1G
APOSOR70056-1GEA 21.9 EUR
APOSOR70056-1G
5-Bromo-2-methoxybenzoic acid
5-Bromo-2-methoxybenzoic acid
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00093713
CAS Number: 2476-35-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR