3-(2,3-Dihydro-1-benzofuran-5-yl)acrylic acid 95+%

Supplier: Apollo Scientific
Warning

Synonyms: (E)-3-(2,3-Dihydrobenzofuran-5-yl)acrylic acid

OR7839-1G
APOSOR7839-1GEA 58 EUR
APOSOR7839-1G
3-(2,3-Dihydro-1-benzofuran-5-yl)acrylic acid 95+%
3-(2,3-Dihydro-1-benzofuran-5-yl)acrylic acid
Formula: C₁₁H₁₀O₃
MW: 190.2 g/mol
Melting Pt: 170…173 °C
Storage Temperature: Ambient
MDL Number: MFCD03002828
CAS Number: 203505-84-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR