1-(3-Nitropyridin-2-yl)piperidine-4-carbonitrile 95+%

Supplier: Apollo Scientific
Warning

OR7937-250MG
APOSOR7937-250MGEA 24.8 EUR
APOSOR7937-250MG
1-(3-Nitropyridin-2-yl)piperidine-4-carbonitrile 95+%
1-(3-Nitropyridin-2-yl)piperidine-4-carbonitrile
Formula: C₁₁H₁₂N₄O₂
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00106383
CAS Number: 671191-99-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR