1-Boc-4-piperidone 98%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Oxo-piperidine-1-carboxylic acidtert-butyl ester, N-BOC-4-Piperidone, N-tert-BOC-4-Piperidinone

OR8050-100G
APOSOR8050-100GEA 94.4 EUR
APOSOR8050-100G
1-Boc-4-piperidone 98%
1-Boc-4-piperidone
Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
Melting Pt: 72…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00151800
CAS Number: 79099-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR