2,8-Dichloroquinoline 98%

Supplier: Apollo Scientific

OR901662-25G
APOSOR901662-25GEA 142 EUR
APOSOR901662-25G
2,8-Dichloroquinoline 98%
2,8-Dichloroquinoline
Formula: C₉H₅Cl₂N
MW: 198.0500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00832077
CAS Number: 4470-83-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR