2-Bromo-4-hydroxypyridine 96%

Supplier: Apollo Scientific

Synonyms: 2-Bromopyridin-4-ol

OR901771-10G OR901771-1G OR901771-5G
APOSOR901771-10GEA 255 EUR
APOSOR901771-10G APOSOR901771-1G APOSOR901771-5G
2-Bromo-4-hydroxypyridine 96%
2-Bromo-4-hydroxypyridine
Formula: C₅H₄BrNO
MW: 174 g/mol
Melting Pt: 171 °C
Storage Temperature: Ambient
MDL Number: MFCD00234040
CAS Number: 36953-40-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR