N-Benzylacetamide 98%

Supplier: Apollo Scientific

Synonyms: Lacosamide EP Impurity G, N-Acetylbenzylamine

OR904376-100G
APOSOR904376-100GEA 186 EUR
APOSOR904376-100G
N-Benzylacetamide 98%
N-Benzylacetamide
Formula: C₉H₁₁NO
MW: 149.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059204
CAS Number: 588-46-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR