2,5-Dichloro-4-methylpyrimidine 96%

Supplier: Apollo Scientific

OR906028-1G
APOSOR906028-1GEA 69.6 EUR
APOSOR906028-1G
2,5-Dichloro-4-methylpyrimidine 96%
2,5-Dichloro-4-methylpyrimidine
Formula: C₅H₄Cl₂N₂
MW: 163.0100 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13193656
CAS Number: 1192064-63-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR