(±)-2-Bromovaleric acid 98%

Supplier: Apollo Scientific

Synonyms: (±)-2-Bromopentanoic acid, 2-Bromovalericacid

OR907076-500G
APOSOR907076-500GEA 155 EUR
APOSOR907076-500G
(±)-2-Bromovaleric acid 98%
(±)-2-Bromovaleric acid
Formula: H₃C(CH₂)₂BrCHCOOH
MW: 181.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004217
CAS Number: 584-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR