4-Methylpyrazole-5-boronic acid, pinacol ester

Supplier: Apollo Scientific
Warning

Synonyms: 4-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

OR908586-250MG OR908586-100MG
APOSOR908586-250MGEA 207 EUR
APOSOR908586-250MG APOSOR908586-100MG
4-Methylpyrazole-5-boronic acid, pinacol ester
4-Methylpyrazole-5-boronic acid, pinacol ester
Formula: C₁₀H₁₇BN₂O₂
MW: 208.065 g/mol
Storage Temperature: Freezer
MDL Number: MFCD18250651
CAS Number: 1440520-87-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR