3-Methylquinoline 95%

Supplier: Apollo Scientific

OR911987-25G
APOSOR911987-25GEA 62.5 EUR
APOSOR911987-25G
3-Methylquinoline 95%
3-Methylquinoline
Formula: C₁₀H₉N
MW: 143.19 g/mol
Melting Pt: 17
Storage Temperature: Ambient
MDL Number: MFCD00014661
CAS Number: 612-58-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR