3-Chloro-5-methyl-1H-pyrazole 95%

Supplier: Apollo Scientific

Synonyms: 5-Chloro-3-methyl-1H-pyrazole

OR917576-1G
APOSOR917576-1GEA 52.2 EUR
APOSOR917576-1G
3-Chloro-5-methyl-1H-pyrazole 95%
3-Chloro-5-methyl-1H-pyrazole
Formula: C₄H₅ClN₂
MW: 116.55 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00233972
CAS Number: 15953-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR