2-Acetylamino-3-cyclopropylpropionic acid 95%

Supplier: Apollo Scientific

OR918453-1G
APOSOR918453-1GEA 435 EUR
APOSOR918453-1G
2-Acetylamino-3-cyclopropylpropionic acid 95%
2-Acetylamino-3-cyclopropylpropionic acid
Formula: C₈H₁₃NO₃
MW: 171.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09878741
CAS Number: 133992-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR