4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) 95%

Supplier: Apollo Scientific

Synonyms: 4-Methoxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Methoxy-2,2,6,6-tetramethyl-1-piperidinyloxy

OR918603-1G OR918603-5G
APOSOR918603-1GEA 62.5 EUR
APOSOR918603-1G APOSOR918603-5G
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) 95%
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-Methoxy-TEMPO, free radical)
Formula: C₁₀H₂₀NO₂
MW: 186.27 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00270334
CAS Number: 95407-69-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR