N-Boc-4-oxo-L-proline 97%

Supplier: Apollo Scientific
Warning

Synonyms: Boc-4-oxo-Pro-OH, (S)-1-Boc-4-oxo-2-pyrrolidinecarboxylic Acid, (S)-4-Oxopyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, (S)-1-(tert-Butoxycarbonyl)-4-oxo-2-pyrrolidinecarboxylic Acid, N-(tert-Butoxycarbonyl)-4-oxo-L-proline

OR9194-1G
APOSOR9194-1GEA 21.9 EUR
APOSOR9194-1G
N-Boc-4-oxo-L-proline 97%
N-Boc-4-oxo-L-proline
Formula: C₁₀H₁₅NO₅
MW: 229.23 g/mol
Melting Pt: 155…156 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01860669
CAS Number: 84348-37-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR